KEY465195496 Key Organics / BIONET

3-[(1Z)-2-amino-1,2-dicyanoeth-1-en-1-yl]-1-phenylurea

  • MDL number MFCD00170981
  • Cas Number : 51802-29-0
  • Molecular Weight : 227.227

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73459

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3W-0628_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart