KEY465195502 Key Organics / BIONET

3-[(1Z)-2-amino-1,2-dicyanoeth-1-en-1-yl]-1-(4-chlorophenyl)urea

  • MDL number MFCD00238955
  • Cas Number : 865658-74-8
  • Molecular Weight : 261.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73467

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3W-0654_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart