KEY465195506 Key Organics / BIONET

4-{[(5Z)-3-methyl-2,4-dioxo-1,3-thiazolidin-5-ylidene]methyl}benzonitrile

  • MDL number MFCD03618061
  • Cas Number : 367476-03-7
  • Molecular Weight : 244.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73478

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3W-0812_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart