KEY465195508 Key Organics / BIONET

1-[3-methanesulfonyl-5-(trifluoromethyl)pyridin-2-yl]ethan-1-one

  • MDL number MFCD22421564
  • Cas Number : 1400872-34-5
  • Molecular Weight : 267.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160 - 162
Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73480

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3W-0815_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart