KEY465195513 Key Organics / BIONET

5-{3-[4-(4-fluorophenyl)piperazin-1-yl]prop-1-yn-1-yl}pyrimidine

  • MDL number MFCD03848482
  • Cas Number : 865658-83-9
  • Molecular Weight : 296.349

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73488

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3W-0824_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart