KEY465195517 Key Organics / BIONET

N-{6-oxo-4H,5H,6H-cyclopenta[b]thiophen-4-yl}-3-(trifluoromethyl)benzamide

  • MDL number MFCD03848486
  • Cas Number : 865658-93-1
  • Molecular Weight : 325.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73501

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3W-0841_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart