KEY465195520 Key Organics / BIONET

N-{6-oxo-4H,5H,6H-cyclopenta[b]thiophen-4-yl}cyclohexanecarboxamide

  • MDL number MFCD03787107
  • Cas Number : 865658-99-7
  • Molecular Weight : 263.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73509

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3W-0851_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart