KEY465195522 Key Organics / BIONET

2-nitro-N-{6-oxo-4H,5H,6H-cyclopenta[b]thiophen-4-yl}benzene-1-sulfonamide

  • MDL number MFCD03787109
  • Cas Number : 865659-01-4
  • Molecular Weight : 338.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73511

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3W-0853_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart