KEY465195529 Key Organics / BIONET

3-amino-6-ethoxy-5-methoxy-2,3-dihydro-1H-inden-1-one hydrochloride

  • MDL number MFCD03787113
  • Cas Number : 1052545-06-8
  • Molecular Weight : 257.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73530

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3W-0880_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart