KEY465195532 Key Organics / BIONET

5-[(3-fluorophenyl)methoxy]-2-(hydroxymethyl)-4H-pyran-4-one

  • MDL number MFCD03848493
  • Cas Number : 865659-14-9
  • Molecular Weight : 250.225

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73538

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3W-0888_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart