KEY465195534 Key Organics / BIONET

N-{pyrrolo[1,2-a]quinolin-6-yl}thiophene-2-sulfonamide

  • MDL number MFCD03848498
  • Cas Number : 865659-19-4
  • Molecular Weight : 328.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73543

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3W-0893_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart