KEY465195535 Key Organics / BIONET

4-methoxy-N-{pyrrolo[1,2-a]quinolin-6-yl}benzene-1-sulfonamide

  • MDL number MFCD03848499
  • Cas Number : 865659-20-7
  • Molecular Weight : 352.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73544

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3W-0894_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart