KEY465195541 Key Organics / BIONET

(E)-N,N-dimethyl-N'-[5-(naphthalene-2-carbonyl)-1,3-thiazol-2-yl]methanimidamide

  • MDL number MFCD04125195
  • Cas Number : 865659-27-4
  • Molecular Weight : 309.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73552

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/3X-0208_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart