KEY465195542 Key Organics / BIONET

1-(4-{[(4-chlorophenyl)methanesulfonyl]methyl}-4-hydroxypiperidin-1-yl)-2,2-dimethylpropan-1-one

  • MDL number MFCD04125198
  • Cas Number : 865659-29-6
  • Molecular Weight : 387.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73555

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3X-0212_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart