KEY465195558 Key Organics / BIONET

1-{4-hydroxy-4-[(4-methoxybenzenesulfonyl)methyl]piperidin-1-yl}-2,2-dimethylpropan-1-one

  • MDL number MFCD04125219
  • Cas Number : 865659-45-6
  • Molecular Weight : 369.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73578

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3X-0245_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart