KEY465195563 Key Organics / BIONET

ethyl 2-{2-[(E)-[(dimethylamino)methylidene]amino]-1,3-thiazol-5-yl}-2-oxoacetate

  • MDL number MFCD04125225
  • Cas Number : 865659-48-9
  • Molecular Weight : 255.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73585

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3X-0256_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart