KEY465195566 Key Organics / BIONET

(E)-N,N-dimethyl-N'-[5-(2,2,2-trifluoroacetyl)-1,3-thiazol-2-yl]methanimidamide

  • MDL number MFCD04125229
  • Cas Number : 865659-50-3
  • Molecular Weight : 251.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73589

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3X-0265_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart