KEY465195570 Key Organics / BIONET

cyclopropyl({4'-methoxy-[1,1'-biphenyl]-4-yl})methanone

  • MDL number MFCD04125234
  • Cas Number : 865659-55-8
  • Molecular Weight : 252.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73594

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3X-0274_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart