KEY465195584 Key Organics / BIONET

N'-[(1E)-(2H-1,3-benzodioxol-5-yl)methylidene]-1-methyl-3-(trifluoromethyl)-1H-thieno[2,3-c]pyrazole-5-carbohydrazide

  • MDL number MFCD04125262
  • Cas Number : 865659-79-6
  • Molecular Weight : 396.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73624

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3X-0324_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart