KEY465195585 Key Organics / BIONET

N-[(ethylcarbamothioyl)amino]-1-methyl-3-(trifluoromethyl)-1H-thieno[2,3-c]pyrazole-5-carboxamide

  • MDL number MFCD04125263
  • Cas Number : 865659-80-9
  • Molecular Weight : 351.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73625

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3X-0325_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart