KEY465195587 Key Organics / BIONET

2-{[4-(trifluoromethyl)-1,3-thiazol-2-yl]amino}-3,4,5,6,7,8-hexahydroquinazolin-4-one

  • MDL number MFCD04125274
  • Cas Number : 865659-90-1
  • Molecular Weight : 316.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73639

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3X-0346_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart