KEY465195588 Key Organics / BIONET

(E)-N'-{5-[5-(2,4-dichlorophenyl)furan-2-carbonyl]-1,3-thiazol-2-yl}-N,N-dimethylmethanimidamide

  • MDL number MFCD04125277
  • Cas Number : 865659-93-4
  • Molecular Weight : 394.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73642

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3X-0352_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart