KEY465195594 Key Organics / BIONET

3-hydroxy-6-(hydroxymethyl)-2-[(morpholin-4-yl)methyl]-4H-pyran-4-one

  • MDL number MFCD00086315
  • Cas Number : 1429-25-0
  • Molecular Weight : 241.243

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73653

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/3X-0382_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart