KEY465195603 Key Organics / BIONET

[(2-chloro-1,3-thiazol-5-yl)methyl](4-phenylbutan-2-yl)amine

  • MDL number MFCD04125309
  • Cas Number : 865660-16-8
  • Molecular Weight : 280.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73703

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3X-0827_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart