KEY465195605 Key Organics / BIONET

methyl 3-amino-2-{[4-(2,6-dimethylphenyl)piperazin-1-yl]methyl}benzoate

  • MDL number MFCD04125314
  • Cas Number : 865660-19-1
  • Molecular Weight : 353.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73708

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3X-0842_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart