KEY465195606 Key Organics / BIONET

methyl 3-amino-2-[(4-phenylpiperazin-1-yl)methyl]benzoate

  • MDL number MFCD04125318
  • Cas Number : 865660-21-5
  • Molecular Weight : 325.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73712

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3X-0846_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart