KEY465195608 Key Organics / BIONET

methyl 6-amino-5-cyano-4-(4-cyanophenyl)-2-(2-methoxy-2-oxoethyl)-4H-pyran-3-carboxylate

  • MDL number MFCD04125335
  • Cas Number : 865660-39-5
  • Molecular Weight : 353.334

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73735

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3X-0881_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart