KEY465195609 Key Organics / BIONET

5-nitro-2-oxa-11-azatricyclo[10.4.0.0^{4,9}]hexadeca-1(16),4(9),5,7,12,14-hexaen-10-one

  • MDL number MFCD04125340
  • Cas Number : 865660-44-2
  • Molecular Weight : 270.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73741

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3X-0895_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart