KEY465195613 Key Organics / BIONET

2-amino-4,6-bis(piperidin-1-yl)pyrimidine-5-carbaldehyde

  • MDL number MFCD04125347
  • Cas Number : 865660-48-6
  • Molecular Weight : 289.383

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73749

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3X-0910_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart