KEY465195618 Key Organics / BIONET

3-(3-chloro-4-methoxyphenyl)-1-methyl-4-(4-methylbenzoyl)pyrrolidine

  • MDL number MFCD04125354
  • Cas Number : 865660-55-5
  • Molecular Weight : 343.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73756

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3X-0919_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart