KEY465195626 Key Organics / BIONET

9-(2,4-dichlorophenyl)-6,7-dimethoxy-1H,3H,4H,9H-furo[3,4-b]quinolin-1-one

  • MDL number MFCD07021377
  • Cas Number : 900014-92-8
  • Molecular Weight : 392.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73768

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3Y-0052_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart