KEY465195635 Key Organics / BIONET

4-[(2E)-2-[(2H-1,3-benzodioxol-4-yl)methylidene]-2-cyanoacetyl]benzonitrile

  • MDL number MFCD07021397
  • Cas Number : 900014-98-4
  • Molecular Weight : 302.289

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73817

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3Y-0846_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart