KEY465195636 Key Organics / BIONET

3-[(4-hydroxy-3-methoxy-5-methylphenyl)methyl]-2,3-dihydro-1H-indol-2-one

  • MDL number MFCD08281853
  • Cas Number : 882747-48-0
  • Molecular Weight : 283.327

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73818

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3Y-0848_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart