KEY465195638 Key Organics / BIONET

5-amino-1-[3-(trifluoromethyl)phenyl]-1H-pyrazole-4-carbonitrile

  • MDL number MFCD07021398
  • Cas Number : 956930-23-7
  • Molecular Weight : 252.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73822

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3Y-0926_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart