KEY465195640 Key Organics / BIONET

(E)-N'-[4-cyano-1-(4-methoxyphenyl)-1H-pyrazol-5-yl]-N,N-dimethylmethanimidamide

  • MDL number MFCD08064741
  • Cas Number : 957042-59-0
  • Molecular Weight : 269.308

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73830

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3Y-0947_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart