KEY465195641 Key Organics / BIONET

(E)-N-[4-cyano-1-(4-methylphenyl)-1H-pyrazol-5-yl]-N'-hydroxymethanimidamide

  • MDL number MFCD08064742
  • Cas Number : 1007014-67-6
  • Molecular Weight : 241.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73831

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3Y-0948_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart