KEY465195644 Key Organics / BIONET

2-[(5-methoxy-1H-indol-3-yl)methylidene]propanedinitrile

  • MDL number MFCD06213890
  • Cas Number : 771574-78-8
  • Molecular Weight : 223.235

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73841

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3Y-5021_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart