KEY465195646 Key Organics / BIONET

N-[2-(3,4-dimethoxyphenyl)ethyl]-2-hydroxy-5-nitrobenzamide

  • MDL number MFCD07021403
  • Cas Number : 521313-28-0
  • Molecular Weight : 346.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73846

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3Y-5042_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart