KEY465195647 Key Organics / BIONET

4-amino-2-[2-(3,4-dimethoxyphenyl)ethyl]-2,3-dihydro-1H-isoindol-1-one

  • MDL number MFCD07021404
  • Cas Number : 900015-11-4
  • Molecular Weight : 312.369

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73847

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3Y-5044_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart