KEY465195650 Key Organics / BIONET

6-amino-3-[2-(3,4-dimethoxyphenyl)ethyl]-3,4-dihydro-2H-1,3-benzoxazine-2,4-dione

  • MDL number MFCD07021407
  • Cas Number : 900015-13-6
  • Molecular Weight : 342.351

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73852

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3Y-5076_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart