KEY465195659 Key Organics / BIONET

methyl 3-{2-[2-(benzyloxy)acetamido]phenoxy}thiophene-2-carboxylate

  • MDL number MFCD07021419
  • Cas Number : 900018-85-1
  • Molecular Weight : 397.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73866

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3Y-5160_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart