KEY465195660 Key Organics / BIONET

1-(4-acetylphenyl)-N-(cyclopropylmethyl)piperidine-4-carboxamide

  • MDL number MFCD07021427
  • Cas Number : 900018-93-1
  • Molecular Weight : 300.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73875

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3Y-5175_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart