KEY465195673 Key Organics / BIONET

3-fluoro-4-(3-fluorophenoxy)aniline

  • MDL number MFCD08687815
  • Cas Number : 937597-93-8
  • Molecular Weight : 221.207

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75
Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73898

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3Y-7016_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart