KEY465195674 Key Organics / BIONET

ethyl 2-{4-bromo-2-[(1E)-(hydroxyimino)methyl]-6-methoxyphenoxy}acetate

  • MDL number MFCD00127397
  • Cas Number : 338416-53-8
  • Molecular Weight : 332.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73929

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4B-031_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart