KEY465195680 Key Organics / BIONET

1-(2-chlorophenyl)-N-methyl-4-nitro-1H-pyrazol-5-amine

  • MDL number MFCD00127404
  • Cas Number : 318238-10-7
  • Molecular Weight : 252.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73938

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4B-045_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart