KEY465195683 Key Organics / BIONET

(E,2Z)-N'-[(4-chlorophenyl)amino]-2-(methylimino)-N,N-dioxo-2-(pyrrolidin-1-yl)ethanimidamide

  • MDL number MFCD01935896
  • Cas Number : 338416-67-4
  • Molecular Weight : 309.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73951

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4B-064_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart