KEY465195697 Key Organics / BIONET

1-(4-methoxybenzenesulfonyl)-N,N-dimethyl-1H-1,2,3,4-tetrazol-5-amine

  • MDL number MFCD00127458
  • Cas Number : 338417-37-1
  • Molecular Weight : 283.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74012

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4C-036_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart