KEY465195700 Key Organics / BIONET

methyl 2-methyl-5-oxo-5H-chromeno[2,3-b]pyridine-3-carboxylate

  • MDL number MFCD00127463
  • Cas Number : 338417-42-8
  • Molecular Weight : 269.256

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74019

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4C-045_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart