KEY465195708 Key Organics / BIONET

(2Z)-3-(4-methoxyphenyl)-2-(nitromethylidene)-1,3-thiazolidin-4-one

  • MDL number MFCD01935900
  • Cas Number : 1164474-29-6
  • Molecular Weight : 266.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74032

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4C-077_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart