KEY465195712 Key Organics / BIONET

3-oxatricyclo[7.3.1.0^{5,13}]trideca-1(12),5(13),6,8,10-pentaen-2-one

  • MDL number MFCD00138509
  • Cas Number : 518-86-5
  • Molecular Weight : 184.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74051

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4C-112_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart